Order Entry
Denmark
ContactUsLinkComponent
Reverse osmosis water purification system, Millipore®
  172-5087
 :  Merck
undefined
Reverse osmosis water purification system, Millipore®
  172-5087
 :  Merck
  • Description:
    Reverse osmosis module
  • Environmentally Preferable:
  • Production flow rate:
    5 L/h
  • Pk:
    1
This Reverse Osmosis membrane removes 95 to 99% of contaminants including ions, particulates, bacteria, and organic molecules (MW >200kDa)
Height: 41 cm