Cell culture flasks, Nunc™ TripleFlask™
About this item
The Nunc™ TripleFlask™ is a 3-layer flask providing three times the growth surface of a T175 flask for the same footprint, saving space in the incubator.
- Recommended working volume is 200 ml
- Promotes monolayer adherent cell culture
- Also available barcoded (*) with large Code 128
- Nunclon Delta Certificate is included in each case for monolayer formation, cloning efficiency, non-cytotoxic, non-pyrogenic, and sterility
- Available in non-treated format
Perform cell culture expansion and experience 500 cm² total growth area within the footprint of a standard flask using Thermo Scientific™ Nunc™ TripleFlask™ Treated Cell Culture Flasks. Total culture area is achieved by way of the three parallel layers and a straight neck design that works effectively within automation systems.
Nunc™ Delta surface treatment offers maximum adhesion for a broad range of cell types. Barcoding option for automation cell culture.
7 Options Available Below
Product Details & Documents
The Nunc™ TripleFlask™ is a 3-layer flask providing three times the growth surface of a T175 flask for the same footprint, saving space in the incubator.
- Recommended working volume is 200 ml
- Promotes monolayer adherent cell culture
- Also available barcoded (*) with large Code 128
- Nunclon Delta Certificate is included in each case for monolayer formation, cloning efficiency, non-cytotoxic, non-pyrogenic, and sterility
- Available in non-treated format
Perform cell culture expansion and experience 500 cm² total growth area within the footprint of a standard flask using Thermo Scientific™ Nunc™ TripleFlask™ Treated Cell Culture Flasks. Total culture area is achieved by way of the three parallel layers and a straight neck design that works effectively within automation systems.
Nunc™ Delta surface treatment offers maximum adhesion for a broad range of cell types. Barcoding option for automation cell culture.
Documents
Product Family Options
Product Information
- Growth areaClosure typeNeckVersionRecommended working volumePackedAvailabilityPrice
- 500 cm²Filter capStraightNon-treated100 - 200 ml2/packLog in for availability1 * 32 Items5.240,00 kr
- 500 cm²Vent/closeStraightNunclon™Δ coated100 - 200 ml4/packLog in for availability1 * 32 Items5.220,00 kr
- 500 cm²Vent/closeStraightNunclon™Δ coated100 - 200 ml1/packLog in for availability1 * 32 Items5.580,00 kr
- 500 cm²Filter capStraightNunclon™Δ coated100 - 200 ml1/packLog in for availability1 * 32 Items5.240,00 kr
- 500* cm²Filter capStraightNunclon™Δ coated100 - 200 ml1/packLog in for availability1 * 32 Items6.320,00 kr
- 500* cm²Filter capStraightNunclon™Δ coated100 - 200 ml4/packLog in for availability1 * 32 Items6.340,00 kr
- 500 cm²Filter capStraightNunclon™Δ coated100 - 200 ml4/packLog in for availability1 * 32 Items5.100,00 kr