VWR® Traceable® Lollipop™, Precision Thermometer
Water-resistant/shockproof thermometer (IP 64) replaces hazardous mercury units and is ideal for wet labs, wash-down areas, outdoors and virtually all field applications.
- Backlit, rotating ½" high LCD digits are easy to view
- °F/°C switchable
Shockproof in drop tests up to 5 feet. Stainless-steel (SS316) probe is resistant to most laboratory chemicals. Wide range covers 99 percent of the lab’s routine temperature measurements. Solid-state construction eliminates dangers inherent in breakable glass and toxic mercury thermometers.
Optional stainless-steel probe with cable features suction cups which allow versatile placement of water-resistant probe.
: Multi-point calibration on an individually-numbered Traceable® Certificate which assures accuracy from our ISO/IEC 17025:2005 (1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement.
: Supplied with Traceable® Certificate, stainless-steel probe, probe cover with pocket clip, and battery.
Frequently Bought Together