Order Entry
Denmark
ContactUsLinkComponent
 
Organic reference standard, Acetaldehyde diethyl acetal, analytical standard, Supelco®
Catalog # 55795-1ML
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Acetaldehyde diethyl acetal, analytical standard, Supelco®
Catalog # 55795-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 mL
  • Standards:
    Analytical standard
  • CAS No.:
    105-57-7
  • Description:
    Acetaldehyde diethyl acetal, ≥98,0% (GC)
  • Environmentally Preferable:
  • Formula:
    C₆H₁₄O₂
  • Description:
    Acetaldehyde diethyl acetal
  • Boiling point:
    102 °C (lit.)

Specifications

About this item

Acetaldehyde diethyl acetal is one of the precursors utilised for the synthesis of fragrances and pharmaceutical products.

Acetaldehyde diethyl acetal may be used as a reference standard for the determination of acetaldehyde diethyl acetal in: wine vinegars by headspace sorptive extraction−thermal desorption−gas chromatography−mass spectrometry (HSSE−TD−GC−MS); distilled spirits by GC with flame ionisation detection (FID) as well as GC-FID-MS; organic red wines derived from Saccharomyces cerevisiae strains by GC-FID as well as HSSE-GC−MS; cognac (brandy) by HS-GC-MS.