Supelco® PESTANAL® Organic Reference Standard, Thiacloprid
Catalog # 37905-100MG-R
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:111988-49-9
- Description:Thiacloprid
- Environmentally Preferable:
- Storage conditions:2...30 °C
- Hazard class description:H301 - H332 - H336 - H351 - H360FD - H410
- Shipping conditions:STD
- MDL No.:MFCD02101042
- M-Clarity quality level:MQ100
- Synonym(s):[3-(6-Chloro-3-pyridinylmethyl)-2-thiazolidinylidene]cyanamide
- Formula:C₁₀H₉ClN₄S
- Molecular weight:252,72
- Flash point:Not applicable
Specifications
About this item
- GHS signal: GHS06, GHS08, GHS09
Caution: P202 - P261 - P264 - P273 - P301 + P310 - P304 + P340 + P312