VWR®, Water bath stabiliser, AQUA RESIST WB
A ready to use, liquid product designed to stabilise and protect water baths, cooling circuits, water circuits and incubators.
- Working temperature up to 70 °C
- Non corrosive for conventional materials
- Good wetting characteristics
- Low foaming
- Blue dye for visualisation
AQUA RESIST WB is characterised by a specific combination of several ingredients. Surfactants cause stabilisation of the water with simultaneous good wetting. Stabilisers and corrosion inhibitors ensure high material compatibility and thus for a wide range of applications even with longer exposure times. The blue colour makes visual inspection of the existing product possible.
- Environmentally Preferable:
Frequently Bought Together