Insulated boxes, TransPorter Blueline
: DELTA T
TransPorter Blueline boxes are robust, stackable, dishwasher safe and sealable.
- With ergonomic handles
- The box is not easy to be damaged
- Multiple boxes can be stacked for transportation
- Suitable for TempShell system
- A seal can be used to prevent unauthorised opening of the transport containers
Optional inclusion of RFID-chip for temperature monitoring and box identification (enquire for details).
Thermal insulation box with thick thermal insulation material. Vacuum insulation and transparent plastic inliner with cavities for cooling elements. There are three different positions in the box fit S 1 L elements, and at the same time, two 1 L elements can be placed at the bottom according to actual needs.
Frequently Bought Together