Order Entry
Denmark
ContactUsLinkComponent
Chart recorder pens
Catalog # 18-1004-23
Supplier:  Cytiva
Chart recorder pens
Catalog # 18-1004-23
Supplier:  Cytiva
Specifications
  • Description:
    Pens, long-nib, red, for use with chart recorder REC-112
  • Environmentally Preferable:
  • Pk:
    5
Specifications
About this item
Pens for use with paper chart recorders.
  • Response time of 0,2 s (2,55 to 97,5% FSD)
  • Fibre tip
Available in blue or red.