Order Entry
Denmark
ContactUsLinkComponent
Anti-KRT5/KRT6A Mouse Monoclonal Antibody [clone: D5N/16B4]
Anti-KRT5/KRT6A Mouse Monoclonal Antibody [clone: D5N/16B4]
Supplier:  IMMUNOLOGIC
Certificates
Anti-KRT5/KRT6A Mouse Monoclonal Antibody [clone: D5N/16B4]
Supplier:  IMMUNOLOGIC

Specifications

  • Catalog No:
  • ILM7773-C1
  • ILM7773-C05
  • ILM7773-C01
  • Antigen symbol:
  • KRT5/KRT6A
  • KRT5/KRT6A
  • KRT5/KRT6A
  • Antigen name:
  • Keratin 5/Keratin 6A
  • Keratin 5/Keratin 6A
  • Keratin 5/Keratin 6A
  • Antigen synonyms:
  • KRT6D|KRT6C|K6D|K6C|CK6C|EBS2|KRT5A|CK6D
  • KRT6D|KRT6C|K6D|K6C|CK6C|EBS2|KRT5A|CK6D
  • KRT6D|KRT6C|K6D|K6C|CK6C|EBS2|KRT5A|CK6D
  • Antibody type:
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Clone:
  • D5N/16B4
  • D5N/16B4
  • D5N/16B4
  • Reactivity:
  • Human
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Mouse
  • Gene ID:
  • 3852, 3853
  • 3852, 3853
  • 3852, 3853
  • Isotype:
  • IgG1
  • IgG1
  • IgG1
  • ImmunoChemistry:
  • Yes
  • Yes
  • Yes
  • Form:
  • Concentrated
  • Concentrated
  • Concentrated
  • Storage temperature:
  • 2°C - 8°C
  • 2°C - 8°C
  • 2°C - 8°C

Specifications

Related Information

Frequently Bought Together