Silver(II) oxide
:1301-96-8
:
:AgO
:MFCD00044285
:7.44
:123.87 g/mol
:>100 °C (dec.)
:5.1 II
:12,08666
:215-098-2
:3085
:GHS05
:GHS03
- Environmentally Preferable:
- Appearance:Grey to black Powder
- Assay:85.5 to 88 % (AG)
Frequently Bought Together