Silver iodide 99%, pure
: Argentous iodide, Silver monoiodide
:7783-96-2
:
:AgI
:MFCD00003412
:1506 °C (760 mmHg)
:557 °C
:5.68 g/cm³
:234.77 g/mol
:232-038-0
:9 III
:12,08659
:3077
:GHS09
:Ambient
- Environmentally Preferable:
- Appearance:Yellow to yellow-green Powder
- Assay:≥98.5 % (ex Ag)
Frequently Bought Together