Order Entry
Denmark
ContactUsLinkComponent
Germanium(IV) oxide 99.999% (trace metals basis), Electronic Grade
Germanium(IV) oxide 99.999% (trace metals basis), Electronic Grade
Certificates
undefined
Germanium(IV) oxide 99.999% (trace metals basis), Electronic Grade
Specifications
  • Environmentally Preferable:
Specifications
Specification Test Results
  • Appearance:
    White Powder
  • Total trace metal impurities:
    ≤10 ppm