Order Entry
Denmark
ContactUsLinkComponent
Benzotriazole
Benzotriazole
Catalog # EHERC10539500
Supplier:  EHRENSTORFER
CAS Number:  
undefined
Benzotriazole
Catalog # EHERC10539500
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,1 g
  • Seq:
    3020
  • Environmentally Preferable:

Specifications

About this item

Synonyms: 1,2,3-Benzotriazole, 1H-1,2,3-Benzotriazole, 1H-Benzotriazole, 1H-Benzo[d][1,2,3]triazole
 :
 :119.13 g/mol
Melting Pt:99
EINECS:202-394-1
UN:2811
 :GHS02
 :GHS07
Storage Temperature:Ambient
CAS Number:95-14-7
Formula:C₆H₅N₃
MDL Number:MFCD00005699
Boiling Pt:350
Density:1.34 g/cm³ (20 °C)
Flash Pt:170 °C (closed cup)
ADR:6.1 III
Merck Index:13,01109