Hexaphenylbenzene ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 3',4',5',6'-Tetraphenyl-o-terphenyl
CAS Number:992-04-1
:
Formula:C₄₂H₃₀
MDL Number:MFCD00003057
Molecular Weight:534.7 g/mol
Melting Pt:>300 °C
Storage Temperature:Ambient
EINECS:213-591-7
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together