Thymol blue ACS, Reag. Ph. Eur. pH-indicator, Supelco®
Supplier: Merck
Certificates
About this item
Synonyms: Thymolsulphonphthalein, 3,3-Bis(4-hydroxy-5-isopropyl-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
CAS Number:76-61-9
:
Formula:C₂₇H₃₀O₅S
MDL Number:MFCD00005869
Molecular Weight:466.6 g/mol
Melting Pt:221
Merck Index:12,09541
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:200-973-3
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Solubility:0.11 g/l (25 °C)
- Melting Point:221 °C (decomposition)
- Molar Mass:466.59 g/mol
- Bulk Density:350 kg/m3
Specification Test Results
Related Information
Certificates
Search by Lot NumberMerck Brighter titration Titripur®
Product Literature
msds_108176
SDS
ProductPage_108176
Product specification
Frequently Bought Together