3-(Dimethylamino)phenol 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: N,N-Dimethyl-3-hydroxyaniline, 3-Dimethylaminophenol
CAS Number:99-07-0
:
Formula:C₈H₁₁NO
MDL Number:MFCD00002264
Boiling Pt:265
Molecular Weight:137.18 g/mol
Melting Pt:80…85 °C
Flash Pt:148 °C
EINECS:202-727-0
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Grey to brown to purple Powder
- Infrared spectrum:Conforms
- GC:≥96.0 %
- Water:≤0.05 % (Coulometric)
- Color scale:≤10 APHA
- Residue after evaporation:≤0.002 %
- Substance darkened by H2SO4:Passes test
- Sulfur compounds (as S):≤0.003 %
Specification Test Results
Frequently Bought Together