1-Chloro-2-ethoxybenzene ≥99.0% (by GC)
: TCI
: 1-Chloro-2-ethoxybenzene, 2-Chlorophenetole
:614-72-2
:
:C₈H₉ClO
:MFCD00058930
:210°C
:156.61 g/mol
:210-392-7
- Environmentally Preferable:
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 99.0 %
Frequently Bought Together