Order Entry
Denmark
ContactUsLinkComponent
5-Cyano-1-pentyne 98%
5-Cyano-1-pentyne 98%
Certificates
undefined
5-Cyano-1-pentyne 98%

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    Clear colorless to amber Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4390 to 1.4410 (20°C, 589 nm)
  • GC:
    ≥97.5 %

Specification Test Results

Related Information

Frequently Bought Together