Aluminium isopropoxide 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Aluminiumtriisopropylate, Aluminiumisopropylate, Aluminiumtriisopropoxide, Aluminium isopropoxide
CAS Number:555-31-7
:
Formula:C₉H₂₁AlO₃
MDL Number:MFCD00008870
Boiling Pt:125…130 °C (38 mmHg)
Molecular Weight:204.25 g/mol
Melting Pt:129…136 °C
ADR:4.1 II
Merck Index:13,00346
EINECS:209-090-8
UN:3181
:GHS02
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White Powder
- Infrared spectrum:Conforms
- Melting point:129 to 136 °C
- Aluminium (Al):12.9 to 13.5 %
Specification Test Results
Frequently Bought Together