Order Entry
Denmark
ContactUsLinkComponent
L-Methionine ≥99%, white crystalline powder cell culture reagent
L-Methionine ≥99%, white crystalline powder cell culture reagent
Supplier:  MP Biomedicals
Certificates
L-Methionine ≥99%, white crystalline powder cell culture reagent
Supplier:  MP Biomedicals

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Identity Test:
    Passes
  • Purity:
    ≥99%
  • FTIR:
    Conforms to Standard
  • pH (1% aq soln):
    5.6-6.1
  • Loss on Drying:
    ≤0.3%
  • Optical Rotation (c=2, 6N HCl):
    +22.4° to 24.7°
  • Residue on Ignition:
    ≤0.4%

Specification Test Results

Frequently Bought Together