Order Entry
Denmark
ContactUsLinkComponent
 
L(+)-Sodium lactate
Catalog # MOLE25860279-25G
Supplier:  Molekula
CAS Number:  
L(+)-Sodium lactate
Catalog # MOLE25860279-25G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Sodium (S)-2-hydroxypropanoate, (S)-(+)-2-Hydroxypropanoic acid sodium salt, (S)-(+)-2-Hydroxypropionic acid sodium salt, Sodium-(S)-lactate, (S)-(+)-Lactic acid sodium salt, L(+)-Lactic acid sodium salt
 :
 :112.06 g/mol
EINECS:212-762-3
CAS Number:867-56-1
Formula:C₃H₅NaO₃
MDL Number:MFCD00066576