Aluminium potassium sulphate dodecahydrate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Aluminium Potassium sulphate, 12-Hydrate, Aluminum potassium sulphate dodecahydrate, Aluminium potassium bis(sulphate) dodecahydrate, Potassium alum, Alum potassium
CAS Number:7784-24-9
:
Formula:AlK(SO₄)₂·12H₂O
MDL Number:MFCD00149143
Density:1.757
Molecular Weight:474.39 g/mol
Melting Pt:92 °C
Merck Index:12,00373
Storage Temperature:Ambient
EINECS:233-141-3
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together