Order Entry
Denmark
ContactUsLinkComponent
 
Oxalic acid dihydrate 99+%, extra pure
 
undefined
Oxalic acid dihydrate 99+%, extra pure

 

  • Environmentally Preferable:

 

 

  • Appearance:
    White Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    98 to 102 °C
  • Sulfated ash:
    ≤0.1 %
  • Titration with KMnO4:
    ≥99.0 %
  • Heavy metals (as Pb):
    ≤10 ppm

 

 

Frequently Bought Together