Hydrochloric acid 2.0 M HCl (2.0N) volumetric solution, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
Synonyms: Hydrochloric acid solution, Muriatic acid
CAS Number:7647-01-0
:
Formula:HCl
MDL Number:MFCD00011324
Density:1.04 g/cm³
Molecular Weight:36.46 g/mol
Storage Temperature:Ambient
EINECS:231-595-7
UN:1789
ADR:8 III
:GHS05
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Factor:0.999 - 1.001
- BAM/EMPA reference material:effective
- BAM/EMPA Certificate Number:effective
- Traceable to NIST SRM:effective
Specification Test Results
Frequently Bought Together