Order Entry
Denmark
ContactUsLinkComponent
 
Benzalkonium chloride for synthesis, Sigma-Aldrich®
 :  Merck
 
Benzalkonium chloride for synthesis, Sigma-Aldrich®
 :  Merck
 

 

  • Environmentally Preferable:

 

 

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    37 - 41 °C
  • Bulk Density:
    800 kg/m3
  • Boiling Point:
    115 °C
  • pH:
    6 - 8 (10 g/l, H2O, 20 °C)
  • Synonyms:
    Benzyldimethylalkylammonium chloride
  • Hill Formula:
    [RN(CH3)2CH2C6H5]Cl

 

Frequently Bought Together