Molecular sieve A4 (0.4 nm, 4 Å), beads Reag. Ph. Eur. 2 mm, Supelco®
Supplier: Merck
Certificates
About this item
Synonyms: Molecular sieve 0.4 nm, Molecular sieves pack 4A, Linde 4A, Sodium aluminium silicate
CAS Number:70955-01-0
:1318-02-1
MDL Number:MFCD03457537
Density:1.1…2.2 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:215-283-8
Limited Time Sale
Select items below are currently on sale
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Solubility:(20 °C) insoluble
- Bulk Density:700 - 750 kg/m3
- Grade:Reag. Ph Eur
- Synonyms:Sodium aluminium silicate
Specification Test Results
Related Information
Certificates
Search by Lot NumberMERC1.05708.9010_TDS_EN ProductPage_105708
Product specification
Merck Quality Materials for Absorption, Adsorption and Filtration
Product Literature
msds_105708
SDS
Frequently Bought Together