Molybdenum ≥99.95% (metals basis), foil, Thickness: 0.025 mm (0.001 in)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7439-98-7
:
Formula:Mo
MDL Number:MFCD00003465
Boiling Pt:3617
Melting Pt:2622
Density:10.28 g/cm³ (20 °C)
Molecular Weight:95.94 g/mol
Merck Index:12,06317
Storage Temperature:Ambient
EINECS:231-107-2
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together