Order Entry
Denmark
ContactUsLinkComponent
 
IGF-1 (human), ELISA kit
  ENZOADI900150
New Product
 :  
IGF-1 (human), ELISA kit
  ENZOADI900150
New Product
 :  

 

  • Pk:
    1 KIT
  • Host:
  • Primary antibody reactivity:
    Human
  • Target protein:
    IGF-1
  • Description:
    IGF-1 (human), ELISA kit
  • Seq:
    02
  • Environmentally Preferable:
  • Sensitivity:
    48,5 pg/ml (range 187 - 6,000 pg/ml)

 

 

Highly sensitive ELISA, measuring as little as 50pg/ml of IGF-1 in just 4 hours.

For the quantitative determination of human IGF-1 in plasma and serum.

The IGF-1 (human), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 4 hours. Absorbance is read at 450 nm. This kit offers ultra-sensitive measurement of as little as 50pg/ml of IGF-1.

 : Contains Microtiter plate, Conjugate, Antibody, Assay buffer 10 concentrate, Standard, Wash buffer concentrate, TMB substrate, Stop solution 2, Neutralizing reagent 2, 2N HCl.