Anti-COX-2 Rabbit Polyclonal Antibody
Supplier: ENZO LIFE SCIENCES
New Product
Certificates
About this item
Host: Rabbit
- For Immunohistochemistry, Immunoprecipitation, Western Blot
Recognizes human, mouse and sheep COX-2. Does not cross-react with COX-1.
Type: Primary
Antigen: COX-2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Specifications
- Catalog No:
- ENZOBMLSA2370025
- ENZOBMLSA2370100
- Antigen symbol:
- COX-2
- COX-2
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Reactivity:
- Human
- Host:
- Rabbit
- Rabbit
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- Form:
- Liquid, in PBS
- Liquid, in PBS
Specifications
Frequently Bought Together