The diffusible free radical gas nitric oxide (NO) affects a variety of physiological functions, and is a key regulator of the cardiovascular, nervous, and immune systems. NO is synthesized in many tissues from L-arginine, oxygen, and NADPH by three known isoforms of a heme-containing flavoprotein termed NO synthase (nNOS/NOS-I, iNOS/NOS-II, and eNOS/NOS-III). nNOS is a constitutively expressed neuronal NOS isoform that exits in its latent form until it is activated by the binding of calmodulin follo wing elevation of intracellular calcium levels. The C-terminus of nNOS contains a conserved serine residue, Ser1417, analogous to Ser1177 of the constitutively expressed endothelial NOS isoform (eNOS). Phosphorylation of Ser1417 is believed to regulate nNOS activation, particularly in glucose-sensing neurons, where inhibition of AMPK pathways by glucose and leptin serve to suppress nNOS activity, whereas activation of AMPK by insulin leads to nNOS activation.
- For Immunocytochemistry, Western Blot
Recognizes human, mouse and rat nNOS.
Type: Primary
Antigen: NNOS
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
- Catalog No:
- ENZOBMLSA2270025
- ENZOBMLSA2270100
- Antigen symbol:
- NNOS
- NNOS
- Antigen name:
- Nitric Oxide Synthase 1 (Neuronal)
- Nitric Oxide Synthase 1 (Neuronal)
- Antigen synonyms:
- bNOS1|Neuronal Nitric Oxide Synthase|NOS Type 1|Brain NOS
- bNOS1|Neuronal Nitric Oxide Synthase|NOS Type 1|Brain NOS
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Reactivity:
- Human
- Host:
- Rabbit
- Rabbit
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes