About this item
Nuclear factor erythroid 2-related factor 2 (Nrf2) is a transcription factor, which regulates the expression of many detoxification and antioxidant enzymes. Nrf2 can potentially play a significant role in adaptive responses to oxidative stress. Nrf2 belongs to the Cap N Collar subfamily of basic leucine zipper (CNC-bZIP) transcription factors. The NF-E2 related family comprises of Nrf1 (TCFII), Nrf2, and Nrf3 and these proteins heterodimerize with Maf proteins to bind to Maf recognition element (MARE). Under normal conditions, Nrf2 is inactive and localized in the cytoplasm through its association with Keap1. When cells are exposed to electrophiles or oxidative stress generating agents cytoplasm localization of Nrf2 is inactivated and Nrf2 translocates into the nucleus and binds to specific DNA sequence elements known as NF-E2 binding site, Maf recognition element (MARE), stress response element or anti-oxidant response element (ARE). Studies have investigated the role of Nrf2 in urinary bladder carcinogenesis, pulmonary fibrosis, and smoke-induced emphysema. A study reported Nrf2-mediated ARE transcription to be a likely target of preventative therapy in neurodegenerative disorders such as Huntington's disease.
- For Western Blot
Recognizes human, mouse, rat, dog, guinea pig and hamster Nrf2.
Type: Primary
Antigen: NRF2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Specifications
- Catalog No:
- ENZOADIKAPTF125F
- ENZOADIKAPTF125D
- Antigen symbol:
- NRF2
- NRF2
- Antigen name:
- Nuclear Respiratory Factor 2
- Nuclear Respiratory Factor 2
- Antigen synonyms:
- NFE2-related factor 2|nuclear factor|HEBP1|erythroid derived 2|NRF2|nuclear factor erythroid-derived 2-like 2|nuclear factor erythroid 2-related factor 2|NF-E2-related factor 2|like 2
- NFE2-related factor 2|nuclear factor|HEBP1|erythroid derived 2|NRF2|nuclear factor erythroid-derived 2-like 2|nuclear factor erythroid 2-related factor 2|NF-E2-related factor 2|like 2
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Reactivity:
- Human
- Host:
- Rabbit
- Rabbit
- Western blot:
- Yes
- Yes
- Form:
- Liquid. In PBS containing 50% glycerol and 0,09% sodium azide.
- Liquid. In PBS containing 50% glycerol and 0,09% sodium azide.
- Purification:
- Protein A affinity purified
- Protein A affinity purified