Dino-Lite models with USB 3.0 offer an image transfer speed of up to 45 fps at a 1280×960 resolution. USB 3.0 adds a new transfer mode called "SuperSpeed" (SS) capable of transferring data up to 5 Gbits/s (625 MB/s). SuperSpeed allows users to take advantage of the full power of the Dino-Lite Edge Series optics including improved colour accuracy and image quality.
- Super speed USB 3.0 connection
- Metal housing: Microscopes are ESD-safe
- Edge series: High-quality optics provide a very sharp, bright and natural colour image with very low aberration and vignetting
- Integrated polariser greatly reduces glare on reflective surfaces (included with all models except AM73115MTF)
- Model AM73115MTF: Extra Long Working Distance (ELWD) of up to 48 cm
Models with Automatic Magnification Reading (AMR): Allows the included software to read the magnification setting from the microscope device, simplifying the measuring process.
Models with Extended Depth of Field (EDOF): Automatically capture and combine multiple images at different focus levels to produce a final image with greater depth of field.
Models with Extended Dynamic Range (EDR): Automatically capture and combine multiple images at different exposure levels to create an image with greater dynamic range.
Models with Flexible LED control (FLC): Additional LED controls within the included software for adjusting LED brightness and on/off toggle by quadrant.