Anti-Desmin Polyclonal Antibody
Catalog # BSBTPB9105
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen symbol:Desmin
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Polyclonal antibody for Desmin/DES detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. Desmin/DES information: Molecular Weight: 53536 MW; Subcellular Localization: Cytoplasm. Cell membrane, sarcolemma .
Type: Primary
Antigen: Desmin
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse; Rat