Sodium hexacyanoferrate(II) decahydrate
: Sodium ferrocyanide decahydrate
:14434-22-1
:
:C₆H₂₀FeN₆Na₄O₁₀
:MFCD00085360
:484.062 g/mol
:Ambient
- Environmentally Preferable:
- Appearance:Yellow Crystalline powder or crystals
- Loss on drying:≤1 % (free moisture)
- Titration:≥98.5 %
- Chloride (Cl):≤0.1 %
- Sulfate (SO4):≤0.07 %
- Insoluble matter:≤0.01 %
Frequently Bought Together