Multi-channel pipettes, mechanical, variable volume, Research® plus (General Lab Product)
Versatile, ergonomic pipette with innovative design, meeting the highest needs in precision and accuracy.
- Extremely lightweight
- Innovative re-designed principle of piston-stroke pipetting, piston made of organic polymer Fortron® with great chemical, thermal and mechanical resistance
- Lowest aspirating, dispensing, blow-out and tip ejection forces
- Spring loaded retractable nose cone to ensure tight seal and prolong nose cone life, as well as exact tip alignment
- Individual channel removal for quick and easy servicing
- Fully autoclavable
Designed and constructed for general laboratory and research applications in Life Science. Typical applications include: Food & beverages, forensic, cosmetics, agriculture, animal studies, chemical analytics, research applications at medical and academic facilities.
: Without CE marking.
- Catalog No:
- 613-6975
- 613-6976
- 613-6977
- 613-6978
- 613-6979
- 613-5905
- 613-5908
- 613-6974
- 613-5903
- 613-5906
- 613-5904
- 613-5907
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
Frequently Bought Together