Order Entry
Denmark
ContactUsLinkComponent
Bovine haemoglobin (from Erythrocytes)
Bovine haemoglobin (from Erythrocytes)
Catalog # 374834-10
CAS Number:  
Bovine haemoglobin (from Erythrocytes)
Catalog # 374834-10
CAS Number:  

Specifications

  • Pk:
    10 g
  • Source:
    Erythrocytes
  • Species:
    Bovine
  • Environmentally Preferable:
  • Storage conditions:
    +2...+8 °C
  • Protein/peptide name:
    Hemoglobin
  • Shipping temperature:
    Ambient

Specifications

About this item

Hemoglobin, bovine erythrocytes is an iron-containing oxygen-transport metalloprotein. Suitable as a substrate for proteases, such as papain, pepsin, and trypsin.

  • Lyophilised form
  • Standard Handling
  • Glass bottles used for packaging