Organic reference standard, Hyaluronidase BRP (FIP), Ph.Eur., RS
Catalog # EDQMH1115000
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:50 mg
- Standards:Reference standard
- CAS No.:9001-54-1
- Description:Hyaluronidase BRP (FIP)
- Environmentally Preferable:
- Formula:C₁₈H₁₄O₇
- Storage temperature:2 to 8°C
- Description:Hyaluronidase BRP (FIP)
Specifications
About this item
- Pharmaceutical applications