Order Entry
Denmark
ContactUsLinkComponent
Anti-ORAI1 Mouse Monoclonal Antibody [clone: 3F6H5 ]
Anti-ORAI1 Mouse Monoclonal Antibody [clone: 3F6H5 ]
  PRSIPM-5205
undefined
Anti-ORAI1 Mouse Monoclonal Antibody [clone: 3F6H5 ]
  PRSIPM-5205

 

  • Antibody type:
    Primary
  • Antigen name:
    ORAI calcium release-activated calcium modulator 1
  • Antigen symbol:
    ORAI1
  • Clonality:
    Monoclonal
  • Clone:
    3F6H5
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Format:
    Purified Antibody
  • Form:
    Liquid
  • Gene ID:
    84876
  • Antigen synonyms:
    ORAI1 Antibody [3F6H5] : IMD9, ORAT1, CRACM1, TMEM142A
  • Molecular weight:
    Predicted: 33 kDa

    Observed: 56 kDa
  • Storage temperature:
    ORAI1 monoclonal antibody can be stored at -20˚C, stable for one year.
  • Concentration:
    1 mg/mL
  • Shipping temperature:
    4°C
  • Immunogen:
    Mouse monoclonal ORAI1 antibody was raised against a 16 amino acid synthetic peptide from near the carboxy terminus of human ORAI1.
  • Purification:
    ORAI1 Monoclonal Antibody is immunoaffinity chromotography purified IgG.
  • Size:
    100 μg
  • Pk:
    100 µG

 

 

ORAI1 Monoclonal Antibody: Antigen stimulation of immune cells triggers Ca++ entry through Ca++ release-activated Ca++ (CRAC) channels. ORAI1 is a recently identified four-transmembrane spanning protein that is an essential component of CRAC. A missense mutation in this protein in humans is the cause of one fo rm of hereditary severe combined immune deficiency (SCID) which results in ablated T-cell Ca++ entry. It has been suggested that ORAI1 functions as a highly selective Ca++ plasma membrane channel that is gated through interactions with STIM1, the store-activated endoplasmic reticulum Ca++ sensor. ORAI1 often migrates at a higher than expected molecular weight in SDS-PAGE. This antibody is predicted to have no cross-reactivity to ORAI2 or ORAI3.

ORAI1 antibody can be used for detection of ORAI1 by Western blot at 1 - 2 μg/mL. Antibody can also be used for immunohistochemistry starting at 2.5 μg/mL. For immunofluorescence start at 20 μg/mL.

Type: Primary
Antigen: ORAI1
Clonality: Monoclonal
Clone: 3F6H5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat