Human recombinant Esterase D (from E. coli)
Catalog # PRSI91-115
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Enzyme type:Recombinant
- Source:E. coli
- Species:Human
- Environmentally Preferable:
- Tag sequence:C-6 His tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
- Enzyme name:Esterase D
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular weight:32.59 kD
- Sequence:Met1-Ala282
- Formulation:Supplied as a 0.2 µm filtered solution of 20 mM TrisHCl, 10% Glycerol, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Specifications
About this item
Human Esterase D is a serine hydrolase that is involved in the detoxification of formaldehyde. Esterase D plays a part in a variety of substrates, including O-acetylated sialic acids, which may involves in the recycling of sialic acids. Esterase D can be used as a genetic marker for retinoblastoma and Wilson’s disease.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.