Order Entry
Denmark
ContactUsLinkComponent
Human recombinant FGF21 (from E. coli)
Human recombinant FGF21 (from E. coli)
Catalog # PRSI91-203
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant FGF21 (from E. coli)
Catalog # PRSI91-203
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Environmentally Preferable:
  • Tag sequence:
    N-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    Fibroblast Growth Factor-21|FGFL
  • Protein/peptide name:
    FGF21
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    21.69 kD
  • Sequence:
    His29-Ser209
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM Tris, 100mM NaCl, 2mM EDTA, pH9.0 . Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Fibroblast Growth Factor 21 (FGF21) is a growth factor that belongs to the FGF family. FGF family proteins play a central role during prenatal development and postnatal growth and regeneration of mamy tissues, by promoting cellular proliferation and differentiation. FGF21 is a potent activator of glucose uptake on adipocytes, protects animal from diet-induced obesity when overexpression in transgenic mice, and lower blood glucose and triglyceride levels when therapeutically adiministered to diabetic redents. FGF21 is produced by hepatocytes in reponse to free fatty acid stimulation of a PPARa/RXR dimeric complex. This situation occurs clinically during starvation, or following the ingestionof a highly-fat/low-carbohydrate diet.Upon FGF21 secretion, white adipose tissue is induced to release free fatty acids from triglyceride stores. Once free fatty acid reach hepatocytes, they are oxidized and reduced to acetyl-CoA. The acetyl-CoA is recombined into 4-carbon ketone bodies, release, and transported to peripheral tissue for TCA processing and energy generation.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: N-6 His tag