Strep-tag II peptide
PRSI4337P
:
- Pk:50 µG
- Source:
- Species:
- Environmentally Preferable:
- Storage conditions:Store at –20 °C, stable for one year.
- Protein/peptide name:Strep-tag II Peptide
- Tested applications:Blocking
Strep-tag II peptide is used for blocking the activity of strep-tag II antibody.