Human GTSP1 peptide (14 aa in the center)
Catalog # PRSI4413P
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Source:
- Species:Human
- Environmentally Preferable:
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:14 aa in the center
- Protein/peptide name:GTSP1 Peptide (14 aa in the center)
- Tested applications:Blocking
Specifications
About this item
GSTP1 peptide is used for blocking the activity of GTSP1 antibody.