Anti-CCL8 Mouse Monoclonal Antibody [clone: 1.1_2D4-1A3]
PRSIXP-5730-M
- Antibody type:Primary
- Antigen name:Chemokine (C-C motif) ligand 8
- Antigen symbol:CCL8
- Clonality:Monoclonal
- Clone:1.1_2D4-1A3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Neutralisation:Yes
- Form:Lyophilized
- Gene ID:6355
- Antigen synonyms:HC14|SCYA8|MCP2|MCP-2|SCYA10
- Storage temperature:Stable at room temperature for at least one month and for greater than a year when kept at –20 °C. Reconstitute in sterile water to a concentration of 0.1 - 1.0 mg/mL. Stable for at least six weeks at 2-4 °C when reconstituted.
- Shipping temperature:RT
- Immunogen:The monoclonal antibody was derived form BALB/c X ICR F1 mice and belongs to the IgG1K isotype.
- Purification:The monoclonal antibody against human MCP-2 designated as VL-1 was purified from ascites fluid on a Protein A affinity column.
- Size:500 µg
- Pk:500 µG
MCP 2 is a chemotactic factor that attracts monocytes, lymphocytes, basophils and eosinophils. MCP 2 is important to inflammatory host responses, and is found in the highest concentration in the small intestine and peripheral blood cells.
Type: Primary
Antigen: CCL8
Clonality: Monoclonal
Clone: 1.1_2D4-1A3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human