Anti-CCL16 Mouse Monoclonal Antibody [clone: 1.2_5G3-1D7]
Catalog # PRSIXP-5722-M
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Chemokine (C-C motif) ligand 16
- Antigen symbol:CCL16
- Clonality:Monoclonal
- Clone:1.2_5G3-1D7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:Lyophilized
- Gene ID:6360
- Antigen synonyms:NCC-4|CCL16|SCYL4|Small inducible cytokine subfamily A (Cys Cys) member 16|Liver expressed chemokine|ILINCK|CKb12|LCC1|Mtn-1|Monotactin 1|HCC4|LEC|C-C motif chemokine 16|Mtn1|SCYA16|NCC4|Small inducible cytokine A16 precursor|HCC-4|Chemokine CC-4|LCC-1|LMC
- Storage temperature:Stable at room temperature for at least one month and for greater than a year when kept at –20 °C. Reconstitute in sterile water to a concentration of 0.1 - 1.0 mg/mL. Stable for at least six weeks at 2-4 °C when reconstituted.
- Shipping temperature:RT
- Immunogen:The monoclonal antibody was produced in BALB/c X ICR F1 mice using recombinant hLEC as the immunizing antigen.
- Purification:This IgG1K antibody was purified from ascites fluid on a Protein A affinity column.
- Size:500 µg
- Pk:500 µG
Specifications
About this item
LEC displays chemotactic activity for lymphocytes and monocytes but not for neutrophils. It also shows a potent myelosuppressive activity and suppresses proliferation of myeloid progenitor cells. Its expression is upregulated by IL10.
Type: Primary
Antigen: CCL16
Clonality: Monoclonal
Clone: 1.2_5G3-1D7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human