Specifikationer
- Antigen name:GLI pathogenesis-related 1
- Antigen symbol:GLIPR1
- Clonality:Polyclonal
- Konjugation:Unconjugated
- ELISA:Yes
- Vært:Rabbit
- ImmunFluorescens:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Antigen synonymer:GLIPR1|RTVP1|Protein RTVP-1|GLIPR|Glioma pathogenesis-related protein 1|CRISP7|GliPR 1
- Koncentration:1 mg/mL
- Immunogen:GLIPR1 antibody was raised against an 18 amino acid peptide near the center of human GLIPR1.
- Størrelse:100 µg
- Pakn:100 µG
Specifikationer
Om denne vare
The glioma pathogensis-related protein 1 (GLIPR1) is similar to both the pathogenesis-related protein (PR) superfamily and the cysteine-rich secretory protein (CRISP) family. GLIPR1 is a tumor suppressor whose expression is regulated by p53; its increased expression is associated with myelomocytic differentiation in macrophages, whereas decreased expression of this gene through gene methylation is associated with prostate cancer. The GLIPR1 gene is part of a p53 target gene cluster that includes GLIPR1L1 and GLIPR1L2, two highly homologous proteins whose expression patterns vary.
GLIPR1 antibody is affinity chromatography purified via peptide column.
GLIPR1 antibody is supplied in PBS containing 0.02% sodium azide.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: GLIPR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse