- Antigen name:KN motif And ankyrin Repeat Domains 2
- Antigen symbol:KANK2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Antigen synonyms:SIP|ANKRD25|KIAA1518|KN motif and ankyrin repeat domain-containing protein 2|MXRA3|Ankyrin repeat domain-containing protein 25|KANK2
- Concentration:1 mg/mL
- Immunogen:KANK2 antibody was raised against a 16 amino acid peptide near the carboxy terminus of human KANK2 .
- Size:100 µg
- Pk:100 µG
KANK2 Antibody: Ankyrins are membrane adaptor molecules that play important roles in the control of cytoskeleton formation by regulating actin polymerization. KANK2 (KN motif and ankyrin repeat domain-containing protein 2), was initially identified as a steroid receptor coactivator (SRC) interacting protein (SIP) that could sequester steroid receptor coactivators in the cytoplasm. More recent experiments have shown that while KANK2 is widely expressed, in kidney podocytes, KANK2 localizes to foot processes, suggesting that KANK2 may contribute to controlling actin dynamics in podocyte foot processes.
KANK2 Antibody is affinity chromatography purified via peptide column.
KANK2 Antibody is supplied in PBS containing 0.02% sodium azide.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: KANK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat