Specifications
- Antigen name:Serum Deprivation Response
- Antigen symbol:SDPR
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Antigen synonyms:cavin-2|SDPR|CAVIN2|SDR|PS-p68|Cavin-2|Serum deprivation-response protein
- Concentration:1 mg/mL
- Immunogen:Rabbit polyclonal SDPR antibody was raised against an 18 amino acid peptide near the amino terminus of human SDPR.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
SDPR Antibody: The serum deprivation-response protein (SDPR) is a calcium-independent phospholipid-binding protein whose expression is increased in serum-starved cells. SDPR is a substrate for protein kinase C (PKC) phosphorylation and recruits the polymerase I and transcript release factor (PTRF) to caveolae. Removal of this protein causes caveolae loss and its over-expression results in caveolae deformation and membrane tubulation. Both SDPR and PTRF, as well as the other member of the cavin family PRKCDBP were down regulated in breast cancer cell lines and breast tumor tissue, suggesting that expression of the cavin family proteins could be a useful prognostic indicator of breast cancer progression.
SDPR Antibody is affinity chromatography purified via peptide column.
SDPR Antibody is supplied in PBS containing 0.02% sodium azide.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: SDPR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human