Anti-CRYM Mouse Monoclonal Antibody
PRSI51-443
- Antibody type:Tag
- Antigen name:Crystallin, Mu
- Antigen symbol:CRYM
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Antigen synonyms:THBP|DFNA40
- Immunogen:CRYM (NP_001879, 215 a.a. ~ 315 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
- Size:50 µg
- Pk:50 µG
CRYM antibody can be used in immunohistochemistry starting at 5 μg/mL.
Protein A Column
PBS, pH 7.2
Store CRYM antibody at -20 °C. Aliquot to avoid freeze/thaw cycles.
Type: Tag
Antigen: CRYM
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: