Anti-GRM7 Rabbit Polyclonal Antibody
PRSI50-112
- Antigen name:Glutamate Receptor, Metabotropic 7
- Antigen symbol:GRM7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Antigen synonyms:MGLU7|PPP1R87|MGLUR7|GLUR7|glutamate metabotropic receptor 7|GPRC1G
- Immunogen:Phosphopeptide corresponding to amino acid residues surrounding phospho-Ser862 of rat mGluR7.
- Size:100 µl
- Pk:100 µl
Affinity Purified
Affinity Purified
100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.
Type:
Antigen: Grm7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat