Anti-EIF3F Rabbit Polyclonal Antibody
Catalog # PRSI49-422
Supplier: ProSci Inc.
Specifications
- Antigen symbol:EIF3F
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:Chicken,Chimpanzee,Human,Bovine,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Antigen synonyms:EIF3S5|eIF3-p47
- Immunogen:EIF3F antibody was raised against an internal peptide of EIF3F (Human) 114-125
- Tested applications:Immunoaffinity Chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
eIF3f, also known as eukaryotic translation initiation factor 3 subunit 5, eIF-3 epsilon, and eIF3 p47 subunit, binds to the 40S ribosome and promotes the binding of methionyl-tRNA and mRNA. EIF3f also associates with the complex p170-eIF3. eIF-3 is composed of at least 12 different subunits, eIF3f is one of these subunits.
0.02 M potassium phosphate, 0.15 M NaCl, pH 7.2, 0.01% sodium azide.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: EIF3F
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Chimpanzee, Chicken, Human, Mouse, Rat